missing translation for 'onlineSavingsMsg'
Learn More

4-Formyl-3-(trifluoromethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog No. AAH2698803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2698803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2698803 Supplier Thermo Scientific Chemicals Supplier No. H2698803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1072944-24-1 | Molecular Formula: C8H6BF3O3 | Molecular Weight: 217.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072944-24-1
Molecular Formula C8H6BF3O3
Molecular Weight (g/mol) 217.938
MDL Number MFCD09037498
InChI Key OQDHNEKEJVPVDS-UHFFFAOYSA-N
Synonym 4-formyl-3-trifluoromethyl phenyl boronic acid, 4-formyl-3-trifluoromethyl phenylboronic acid, 4-formyl-3-trifluoromethyl benzeneboronic acid, 5-borono-2-formylbenzotrifluoride, 4-borono-2-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-formylphenylboronic acid, acmc-2098oi, 4-formyl-3-trifluoromethyl phenyl boronicacid
PubChem CID 44755197
IUPAC Name [4-formyl-3-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C=O)C(F)(F)F)(O)O

Specifications

Melting Point 200°C to 202°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 217.94
Percent Purity 95%
Chemical Name or Material 4-Formyl-3-(trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only