CAS RN 1072944-24-1
IUPAC Name:
[4-formyl-3-(trifluoromethyl)phenyl]boronic acid
Synonyms:
4-formyl-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
217.94
Molecular Formula:
C8H6BF3O3
InChi Key:
OQDHNEKEJVPVDS-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=C(C=O)C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4-Formyl-3-(trifluoromethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
4-Formyl-3-(trifluoromethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
CAS: 1072944-24-1 Molecular Formula: C8H6BF3O3 Molecular Weight (g/mol): 217.938 MDL Number: MFCD09037498 InChI Key: OQDHNEKEJVPVDS-UHFFFAOYSA-N Synonym: 4-formyl-3-trifluoromethyl phenyl boronic acid,4-formyl-3-trifluoromethyl phenylboronic acid,4-formyl-3-trifluoromethyl benzeneboronic acid,5-borono-2-formylbenzotrifluoride,4-borono-2-trifluoromethyl benzaldehyde,3-trifluoromethyl-4-formylphenylboronic acid,acmc-2098oi,4-formyl-3-trifluoromethyl phenyl boronicacid PubChem CID: 44755197 IUPAC Name: [4-formyl-3-(trifluoromethyl)phenyl]boronic acid SMILES: B(C1=CC(=C(C=C1)C=O)C(F)(F)F)(O)O