missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid, 98%
CAS: 188715-40-4 | C23H25NO4 | 379.46 g/mol
$157.82 - $575.49
Chemical Identifiers
| CAS | 188715-40-4 |
|---|---|
| Molecular Formula | C23H25NO4 |
| Molecular Weight (g/mol) | 379.46 |
| MDL Number | MFCD01862347,MFCD00273480 |
| InChI Key | MLMIBGARTUSGND-UHFFFAOYSA-N |
| Synonym | fmoc-4-amc-oh, fmoc-tranexamic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, n-fmoc-tranexamic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid |
| PubChem CID | 2756125 |
| IUPAC Name | 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid |
| SMILES | OC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6626603
|
Thermo Scientific Chemicals
H6626603 |
1 g |
Each for $157.82
|
|
|||||
|
AAH6626606
|
Thermo Scientific Chemicals
H6626606 |
5 g |
Each for $575.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 188715-40-4 | |
| 379.46 | |
| MLMIBGARTUSGND-UHFFFAOYSA-N | |
| 2756125 | |
| OC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1 |
| C23H25NO4 | |
| MFCD01862347,MFCD00273480 | |
| fmoc-4-amc-oh, fmoc-tranexamic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, n-fmoc-tranexamic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid | |
| 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid |
Specifications
| 188715-40-4 | |
| C23H25NO4 | |
| 1 g | |
| fmoc-4-amc-oh, fmoc-tranexamic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid, n-fmoc-tranexamic acid, trans-4-fmoc-aminomethyl cyclohexanecarboxylic acid, fmoc-trans-4-aminomethyl-cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-ylmethoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 1r,4r-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexane-1-carboxylic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino-methyl cyclohexanecarboxylic acid, trans-4-9h-fluoren-9-yl methoxy carbonyl amino methyl cyclohexanecarboxylic acid | |
| OC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1 | |
| 379.46 | |
| 379.18 | |
| Powder |
| White | |
| MFCD01862347,MFCD00273480 | |
| 7831105 | |
| MLMIBGARTUSGND-UHFFFAOYSA-N | |
| 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid | |
| 2756125 | |
| 98% | |
| 4-(Fmoc-aminomethyl)cyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only