accessibility menu, dialog, popup

UserName

CAS RN 188715-40-4

IUPAC Name: 4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]cyclohexane-1-carboxylic acid
Synonyms: 4-({[(9H-fluoren-9-ylmethoxy)carbonyl]amino}methyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 379.46
Molecular Formula: C23H25NO4
InChi Key: MLMIBGARTUSGND-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(CNC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results