missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Fmoc-amino)butyric acid, 95%
CAS: 116821-47-7 | C19H19NO4 | 325.364 g/mol
$148.45 - $513.70
Chemical Identifiers
| CAS | 116821-47-7 |
|---|---|
| Molecular Formula | C19H19NO4 |
| Molecular Weight (g/mol) | 325.364 |
| MDL Number | MFCD00144889 |
| InChI Key | ACUIFAAXWDLLTR-UHFFFAOYSA-N |
| Synonym | fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid |
| PubChem CID | 2756086 |
| IUPAC Name | 4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6614706
|
Thermo Scientific Chemicals
H6614706 |
5 g |
Each for $148.45
|
|
|||||
|
AAH6614714
|
Thermo Scientific Chemicals
H6614714 |
25 g |
Each for $513.70
|
|
|||||
Description
4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Fmoc-amino)butyric acid is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 116821-47-7 | |
| 325.364 | |
| ACUIFAAXWDLLTR-UHFFFAOYSA-N | |
| 2756086 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O |
| C19H19NO4 | |
| MFCD00144889 | |
| fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid | |
| 4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid |
Specifications
| 116821-47-7 | |
| C19H19NO4 | |
| MFCD00144889 | |
| 7491485 | |
| Slightly soluble in water. | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O | |
| 325.364 | |
| 325.36 | |
| Powder |
| White | |
| FmocNHCH2CH2CH2CO2H | |
| 5 g | |
| fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid | |
| ACUIFAAXWDLLTR-UHFFFAOYSA-N | |
| 4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid | |
| 2756086 | |
| 95% | |
| 4-(Fmoc-amino)butyric acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only