missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorostyrene (stabilized with TBC) 98.0+%, TCI America™

Catalog No. F04615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04615G 5 g
F046125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04615G Supplier TCI America Supplier No. F04615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 405-99-2
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.142
MDL Number MFCD00000361
InChI Key JWVTWJNGILGLAT-UHFFFAOYSA-N
Synonym 4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp
PubChem CID 67883
IUPAC Name 1-ethenyl-4-fluorobenzene
SMILES C=CC1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 g
UN Number 1993
Formula Weight 122.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorostyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator