accessibility menu, dialog, popup

UserName

CAS RN 405-99-2

IUPAC Name: 1-ethenyl-4-fluorobenzene
Synonyms: benzene, 1-ethenyl-4-fluoro-
Molecular Weight (g/mol): 122.14
Molecular Formula: C8H7F
InChi Key: JWVTWJNGILGLAT-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C)C=C1
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (4)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (6)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)