missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorostyrene, 97%, stabilized

CAS: 405-99-2 | C8H7F | 122.14 g/mol

$75.49 - $247.50

Chemical Identifiers

CAS 405-99-2
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.14
MDL Number MFCD00000361
InChI Key JWVTWJNGILGLAT-UHFFFAOYSA-N
Synonym 4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp
PubChem CID 67883
IUPAC Name 1-ethenyl-4-fluorobenzene
SMILES C=CC1=CC=C(C=C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC168340010
SDS View Documents
Thermo Scientific Chemicals
168340010
1 g Glass bottle N/A
Only null left
 
AC168340100
SDS View Documents
Thermo Scientific Chemicals
168340100
10 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

405-99-2
122.14
JWVTWJNGILGLAT-UHFFFAOYSA-N
67883
C=CC1=CC=C(C=C1)F
C8H7F
MFCD00000361
4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp
1-ethenyl-4-fluorobenzene

Specifications

405-99-2,98-29-3
100.0
1.0240g/mL
26°C
96% min. (GC)
C8H7F
H2C=CHC6H4F
1 g
4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp
C=CC1=CC=C(C=C1)F
122.14
67883
97%
4-Fluorostyrene, Stabilized
96.0
Colorless to Yellow
145°C to 148°C
Authentic
Glass bottle
1.5140 to 1.516
MFCD00000361
1.024
JWVTWJNGILGLAT-UHFFFAOYSA-N
1-ethenyl-4-fluorobenzene
0.1% tert.-butylcatechol
122.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-975-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only