Learn More
4-Fluorostyrene, 97+%, stab. with 0.1% 4-tert-butylcatechol
CAS: 405-99-2 | C8H7F | 122.142 g/mol
Supplier: Thermo Scientific Chemicals A1477514
Description
4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorostyrene, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from acids, bases, oxidizing agents, halogens.
Specifications
| 4-Fluorostyrene | |
| 405-99-2 | |
| 1.024 | |
| 26°C (78°F) | |
| C8H7F | |
| MFCD00000361 | |
| UN1993 | |
| 4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp | |
| JWVTWJNGILGLAT-UHFFFAOYSA-N | |
| 1-ethenyl-4-fluorobenzene | |
| 67883 | |
| ≥97% |
| stabilized with 0.1% 4-tert-butylcatechol | |
| -36°C | |
| 145°C to 148°C | |
| Recognizable | |
| 1.515 | |
| 25 g | |
| 1634203 | |
| Insoluble in water. | |
| C=CC1=CC=C(C=C1)F | |
| 122.142 | |
| 122.14 |
Chemical Identifiers
| 405-99-2 | |
| 122.142 | |
| JWVTWJNGILGLAT-UHFFFAOYSA-N | |
| 67883 | |
| C=CC1=CC=C(C=C1)F |
| C8H7F | |
| MFCD00000361 | |
| 4-fluorostyrene, 1-fluoro-4-vinylbenzene, p-fluorostyrene, benzene, 1-ethenyl-4-fluoro, styrene, p-fluoro, para-fluorostyrene, 1-fluoro-4-vinyl-benzene, 4-fluoro-styrene, acmc-1cszp | |
| 1-ethenyl-4-fluorobenzene |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P380-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 206-975-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only