missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorosalicylic Acid 98.0+%, TCI America™

Catalog No. p-7129800
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
F06371G 1 g
F06375G 5 g
F063725G 25 g
3 options

Catalog No. F06371G

Supplier: TCI America F06371G

Only null left

Chemical Identifiers

CAS 345-29-9
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD00153168
InChI Key TTZOLDXHOCCNMF-UHFFFAOYSA-N
Synonym 4-fluorosalicylic acid, 4-fluorosalicyclic acid, 2-hydroxy-4-fluorobenzoic acid, 4-fluoro-2-hydroxy-benzoic acid, fluorohydroxybenzoicacid5, benzoic acid, 4-fluoro-2-hydroxy, 4-fluorosalicylicacid, zlchem 109, pubchem3493
PubChem CID 67661
IUPAC Name 4-fluoro-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(F)C=C1O

Specifications

Melting Point 189°C
Quantity 1 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorosalicylic Acid