accessibility menu, dialog, popup

UserName

CAS RN 345-29-9

IUPAC Name: 4-fluoro-2-hydroxybenzoic acid
Synonyms: 4-fluoro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 156.11
Molecular Formula: C7H5FO3
InChi Key: TTZOLDXHOCCNMF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(F)C=C1O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)