missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorosalicylic Acid 98.0+%, TCI America™

Catalog No. F06375G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06375G 5 g
F06371G 1 g
F063725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F06375G Supplier TCI America Supplier No. F06375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 345-29-9
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD00153168
InChI Key TTZOLDXHOCCNMF-UHFFFAOYSA-N
Synonym 4-fluorosalicylic acid, 4-fluorosalicyclic acid, 2-hydroxy-4-fluorobenzoic acid, 4-fluoro-2-hydroxy-benzoic acid, fluorohydroxybenzoicacid5, benzoic acid, 4-fluoro-2-hydroxy, 4-fluorosalicylicacid, zlchem 109, pubchem3493
PubChem CID 67661
IUPAC Name 4-fluoro-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(F)C=C1O

Specifications

Melting Point 189°C
Quantity 5 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorosalicylic Acid