missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorosalicylic acid, 98%

Catalog No. p-7027924
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2499203 1 g
AAB2499206 5 g
AAB2499214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2499203 Supplier Thermo Scientific Chemicals Supplier No. B2499203

CAS: 345-29-9 | C7H5FO3 | 156.11 g/mol

4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 345-29-9
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD00153168
InChI Key TTZOLDXHOCCNMF-UHFFFAOYSA-N
Synonym 4-fluorosalicylic acid, 4-fluorosalicyclic acid, 2-hydroxy-4-fluorobenzoic acid, 4-fluoro-2-hydroxy-benzoic acid, fluorohydroxybenzoicacid5, benzoic acid, 4-fluoro-2-hydroxy, 4-fluorosalicylicacid, zlchem 109, pubchem3493
PubChem CID 67661
IUPAC Name 4-fluoro-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(F)C=C1O

Specifications

Melting Point 186°C to 190°C
Quantity 1 g
Beilstein 2209124
Solubility Information Soluble in methanol.
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 4-Fluorosalicylic acid
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-459-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only