Learn More
4-Fluorophenylsulfonylacetonitrile, 97%
CAS: 32083-66-2 | C8H6FNO2S | 199.199 g/mol
Supplier: Thermo Scientific Chemicals A1991606
Description
4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
| 4-Fluorophenylsulfonylacetonitrile | |
| 89°C to 93°C | |
| MFCD00179339 | |
| UN3439 | |
| WBXJZTLPEMITJL-UHFFFAOYSA-N | |
| 2-(4-fluorophenyl)sulfonylacetonitrile | |
| 565732 | |
| 97% |
| 32083-66-2 | |
| C8H6FNO2S | |
| 5 g | |
| 4-fluorobenzenesulfonyl acetonitrile, 2-4-fluorophenyl sulfonyl acetonitrile, 4-fluorobenzenesulphonyl acetonitrile, 4-fluorophenyl sulfonyl acetonitrile, 2-4-fluorobenzenesulfonyl acetonitrile, 4-fluorophenylsulfonylacetonitrile, 4-fluorobenzenesulfonylacetonitrile, 2-4-fluorophenyl sulfonylacetonitrile, 4-fluorophenylsulfonyl acetonitrile | |
| C1=CC(=CC=C1F)S(=O)(=O)CC#N | |
| 199.199 | |
| 199.2 |
Chemical Identifiers
| 32083-66-2 | |
| 199.199 | |
| WBXJZTLPEMITJL-UHFFFAOYSA-N | |
| 565732 | |
| C1=CC(=CC=C1F)S(=O)(=O)CC#N |
| C8H6FNO2S | |
| MFCD00179339 | |
| 4-fluorobenzenesulfonyl acetonitrile, 2-4-fluorophenyl sulfonyl acetonitrile, 4-fluorobenzenesulphonyl acetonitrile, 4-fluorophenyl sulfonyl acetonitrile, 2-4-fluorobenzenesulfonyl acetonitrile, 4-fluorophenylsulfonylacetonitrile, 4-fluorobenzenesulfonylacetonitrile, 2-4-fluorophenyl sulfonylacetonitrile, 4-fluorophenylsulfonyl acetonitrile | |
| 2-(4-fluorophenyl)sulfonylacetonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only