missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylsulfonylacetonitrile, 97%

Catalog No. p-7024730
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1991603 1 g
AAA1991606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1991603 Supplier Thermo Scientific Chemicals Supplier No. A1991603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32083-66-2 | C8H6FNO2S | 199.199 g/mol

4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorophenylsulfonylacetonitrile is used to produce 3-(4-fluoro-benzenesulfonyl)-thiophen-2-ylamine with [1,4]dithiane-2,5-diol. It is also used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 32083-66-2
Molecular Formula C8H6FNO2S
Molecular Weight (g/mol) 199.199
MDL Number MFCD00179339
InChI Key WBXJZTLPEMITJL-UHFFFAOYSA-N
Synonym 4-fluorobenzenesulfonyl acetonitrile, 2-4-fluorophenyl sulfonyl acetonitrile, 4-fluorobenzenesulphonyl acetonitrile, 4-fluorophenyl sulfonyl acetonitrile, 2-4-fluorobenzenesulfonyl acetonitrile, 4-fluorophenylsulfonylacetonitrile, 4-fluorobenzenesulfonylacetonitrile, 2-4-fluorophenyl sulfonylacetonitrile, 4-fluorophenylsulfonyl acetonitrile
PubChem CID 565732
IUPAC Name 2-(4-fluorophenyl)sulfonylacetonitrile
SMILES C1=CC(=CC=C1F)S(=O)(=O)CC#N

Specifications

Melting Point 89°C to 93°C
Quantity 1 g
UN Number UN3439
Formula Weight 199.2
Percent Purity 97%
Chemical Name or Material 4-Fluorophenylsulfonylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only