missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7129538
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F02065G 5 g
F020625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F02065G Supplier TCI America Supplier No. F02065G

Chemical Identifiers

CAS 405-50-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004343
InChI Key MGKPFALCNDRSQD-UHFFFAOYSA-N
Synonym 4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
PubChem CID 9837
IUPAC Name 2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)F

Specifications

Melting Point 85°C
Color White
Boiling Point 164°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorophenylacetic Acid