missing translation for 'onlineSavingsMsg'
Learn More

4-Acide fluorophénylacétique 98,0+%, TCI America™

Numéro de catalogue. F02065G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
F02065G 5 g
F020625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. F02065G Fournisseur TCI America Code fournisseur F02065G

Identifiants chimiques

CAS 405-50-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004343
InChI Key MGKPFALCNDRSQD-UHFFFAOYSA-N
Synonym 4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
PubChem CID 9837
IUPAC Name Acide 2-(4-fluorophényl)acétique
SMILES C1=CC(=CC=C1CC(=O)O)F

Spécifications

Melting Point 85°C
Color Blanc
Boiling Point 164°C
Quantity 5 g
Formula Weight 154.14
Percent Purity ≥98.0% (T)
Physical Form Poudre cristalline
Chemical Name or Material 4-Fluorophenylacetic Acid