missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl sulfone, 99%

CAS: 383-29-9 | C12H8F2O2S | 254.25 g/mol

$186.85 - $186.85

Chemical Identifiers

CAS 383-29-9
Molecular Formula C12H8F2O2S
Molecular Weight (g/mol) 254.25
MDL Number MFCD00000350
InChI Key PLVUIVUKKJTSDM-UHFFFAOYSA-N
Synonym 4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone
PubChem CID 67842
IUPAC Name 1-fluoro-4-(4-fluorophenyl)sulfonylbenzene
SMILES FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119620250
SDS View Documents
Thermo Scientific Chemicals
119620250
25 g Glass bottle
Each for $186.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

383-29-9
254.25
PLVUIVUKKJTSDM-UHFFFAOYSA-N
67842
FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
C12H8F2O2S
MFCD00000350
4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone
1-fluoro-4-(4-fluorophenyl)sulfonylbenzene

Specifications

383-29-9
100.0
Brown to White
99%
C12H8F2O2S
MFCD00000350
4-fluorophenyl sulfone, bis p-fluorophenyl sulfone, 4,4'-sulfonylbis fluorobenzene, bis 4-fluorophenyl sulfone, 4,4'-difluorodiphenyl sulfone, 4-fluorophenyl sulphone, 4-fluorophenylsulfone, benzene, 1,1'-sulfonylbis 4-fluoro, 4,4'-difluorodiphenyl sulphone
FC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(F)C=C1
254.25
254.25
Crystals or Powder
98.5
97.0°C to 100.0°C
Authentic
Glass bottle
(FC6H4)2SO2
25 g
PLVUIVUKKJTSDM-UHFFFAOYSA-N
1-fluoro-4-(4-fluorophenyl)sulfonylbenzene
67842
99%
4-Fluorophenyl sulfone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-847-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only