missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenetole 98.0+%, TCI America™

Catalog No. F06555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06555G Supplier TCI America Supplier No. F06555G
Only null left

Chemical Identifiers

CAS 459-26-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.157
MDL Number MFCD00017953
InChI Key PURLWQWDGIIYBG-UHFFFAOYSA-N
Synonym 1-Ethoxy-4-fluorobenzene
PubChem CID 68018
IUPAC Name 1-ethoxy-4-fluorobenzene
SMILES CCOC1=CC=C(C=C1)F

Specifications

Melting Point -9°C
Color Colorless
Boiling Point 173°C
Quantity 5 g
Formula Weight 140.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorophenetole