accessibility menu, dialog, popup

UserName

CAS RN 459-26-7

CH3OF

CAS RN 459-26-7

IUPAC Name: 1-ethoxy-4-fluorobenzene
Synonyms: benzene, 1-ethoxy-4-fluoro-
Molecular Weight (g/mol): 140.16
Molecular Formula: C8H9FO
InChi Key: PURLWQWDGIIYBG-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results