missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorohippuric Acid 98.0+%, TCI America™

Catalog No. F1162200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1162200MG 200 mg
F11621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1162200MG Supplier TCI America Supplier No. F1162200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 366-79-0
Molecular Formula C9H7FNO3
Molecular Weight (g/mol) 196.16
MDL Number MFCD00462263
InChI Key NVWXSGQHHUSSOU-UHFFFAOYSA-M
Synonym N-(4-Fluorobenzoyl)glycine, 2-(4-Fluorobenzamido)acetic Acid
PubChem CID 699341
IUPAC Name 2-[(4-fluorophenyl)formamido]acetate
SMILES [O-]C(=O)CNC(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 200 mg
Formula Weight 197.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorohippuric Acid