accessibility menu, dialog, popup

UserName

OONHOF

CAS RN 366-79-0

IUPAC Name: 2-[(4-fluorophenyl)formamido]acetate
Synonyms: 2-[(4-fluorophenyl)formamido]acetate
Molecular Weight (g/mol): 196.16
Molecular Formula: C9H7FNO3
InChi Key: NVWXSGQHHUSSOU-UHFFFAOYSA-M
SMILES: [O-]C(=O)CNC(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results