missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorocatechol 98.0+%, TCI America™
$446.64 - $1495.85
Chemical Identifiers
| CAS | 367-32-8 |
|---|---|
| Molecular Formula | C6H5FO2 |
| Molecular Weight (g/mol) | 128.102 |
| MDL Number | MFCD01317539 |
| InChI Key | NFWGQJUHSAGJBE-UHFFFAOYSA-N |
| Synonym | 4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol |
| PubChem CID | 160458 |
| ChEBI | CHEBI:80515 |
| IUPAC Name | 4-fluorobenzene-1,2-diol |
| SMILES | C1=CC(=C(C=C1F)O)O |
Chemical Identifiers
| 367-32-8 | |
| 128.102 | |
| NFWGQJUHSAGJBE-UHFFFAOYSA-N | |
| 160458 | |
| 4-fluorobenzene-1,2-diol |
| C6H5FO2 | |
| MFCD01317539 | |
| 4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol | |
| CHEBI:80515 | |
| C1=CC(=C(C=C1F)O)O |
Specifications
| 367-32-8 | |
| White-Yellow | |
| MFCD01317539 | |
| 1759 | |
| NFWGQJUHSAGJBE-UHFFFAOYSA-N | |
| 4-fluorobenzene-1,2-diol | |
| 160458 | |
| 128.10 | |
| Crystalline Powder |
| 91°C | |
| C6H5FO2 | |
| 1 g | |
| 4-Fluoro-1,2-dihydroxybenzene, 4-Fluoropyrocatechol | |
| C1=CC(=C(C=C1F)O)O | |
| 128.102 | |
| CHEBI:80515 | |
| ≥98.0% (GC) | |
| 4-Fluorocatechol |
Safety and Handling
TSCA : No