accessibility menu, dialog, popup

UserName

OHFHO

CAS RN 367-32-8

IUPAC Name: 4-fluorobenzene-1,2-diol
Synonyms: 4-fluorocatechol
Molecular Weight (g/mol): 128.1
Molecular Formula: C6H5FO2
InChi Key: NFWGQJUHSAGJBE-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results