missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzyl Bromide 97.0+%, TCI America™
$53.90 - $148.25
Chemical Identifiers
| CAS | 459-46-1 |
|---|---|
| Molecular Formula | C7H6BrF |
| Molecular Weight (g/mol) | 189.03 |
| MDL Number | MFCD00000359 |
| InChI Key | NVNPLEPBDPJYRZ-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide |
| PubChem CID | 68021 |
| IUPAC Name | 1-(bromomethyl)-4-fluorobenzene |
| SMILES | FC1=CC=C(CBr)C=C1 |
Chemical Identifiers
| 459-46-1 | |
| 189.03 | |
| NVNPLEPBDPJYRZ-UHFFFAOYSA-N | |
| 68021 | |
| FC1=CC=C(CBr)C=C1 |
| C7H6BrF | |
| MFCD00000359 | |
| 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide | |
| 1-(bromomethyl)-4-fluorobenzene |
Specifications
| 459-46-1 | |
| 177°C | |
| MFCD00000359 | |
| 3265 | |
| NVNPLEPBDPJYRZ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-fluorobenzene | |
| 68021 | |
| ≥97.0% (GC) | |
| 4-Fluorobenzyl Bromide |
| Yellow | |
| C7H6BrF | |
| 5 g | |
| 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide | |
| FC1=CC=C(CBr)C=C1 | |
| 189.03 | |
| 189.03 | |
| Liquid |
Safety and Handling
TSCA : Yes