missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzyl Bromide 97.0+%, TCI America™
Supplier: TCI America F021425G
Specifications
| 4-Fluorobenzyl Bromide | |
| Yellow | |
| C7H6BrF | |
| 25 g | |
| 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide | |
| FC1=CC=C(CBr)C=C1 | |
| 189.03 | |
| 189.03 | |
| Liquid |
| 459-46-1 | |
| 177°C | |
| MFCD00000359 | |
| 3265 | |
| NVNPLEPBDPJYRZ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-fluorobenzene | |
| 68021 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 459-46-1 | |
| 189.03 | |
| NVNPLEPBDPJYRZ-UHFFFAOYSA-N | |
| 68021 | |
| FC1=CC=C(CBr)C=C1 |
| C7H6BrF | |
| MFCD00000359 | |
| 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide | |
| 1-(bromomethyl)-4-fluorobenzene |
Safety and Handling
TSCA : Yes