missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzotrifluoride 98.0+%, TCI America™
$61.61 - $209.84
Chemical Identifiers
| CAS | 402-44-8 |
|---|---|
| Molecular Formula | C7H4F4 |
| Molecular Weight (g/mol) | 164.103 |
| MDL Number | MFCD00000399 |
| InChI Key | UNNNAIWPDLRVRN-UHFFFAOYSA-N |
| Synonym | 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene |
| PubChem CID | 67873 |
| IUPAC Name | 1-fluoro-4-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC=C1C(F)(F)F)F |
Chemical Identifiers
| 402-44-8 | |
| 164.103 | |
| UNNNAIWPDLRVRN-UHFFFAOYSA-N | |
| 67873 | |
| C1=CC(=CC=C1C(F)(F)F)F |
| C7H4F4 | |
| MFCD00000399 | |
| 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene | |
| 1-fluoro-4-(trifluoromethyl)benzene |
Specifications
| 402-44-8 | |
| 103°C | |
| MFCD00000399 | |
| 1993 | |
| UNNNAIWPDLRVRN-UHFFFAOYSA-N | |
| 1-fluoro-4-(trifluoromethyl)benzene | |
| 67873 | |
| ≥98.0% (GC) | |
| 4-Fluorobenzotrifluoride |
| Colorless | |
| C7H4F4 | |
| 5 g | |
| 4-fluorobenzotrifluoride, 1-fluoro-4-trifluoromethyl benzene, p-fluorobenzotrifluoride, benzene, 1-fluoro-4-trifluoromethyl, alpha,alpha,alpha,4-tetrafluorotoluene, p-fluorotrifluoromethylbenzene, p-trifluoromethyl fluorobenzene, 4-flourobenzotrifluoride, a,a,a,4-tetrafluorotoluene, 4-trifluoromethyl fluorobenzene | |
| C1=CC(=CC=C1C(F)(F)F)F | |
| 164.103 | |
| 164.10 | |
| Liquid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator