missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzotrifluoride 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 103°C |
| Quantity | 25 g |
| UN Number | 1993 |
| Formula Weight | 164.10 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Fluorobenzotrifluoride |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Refrigerator |