missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzonitrile 99.0+%, TCI America™
Supplier: TCI America F0221100G
Specifications
| 4-Fluorobenzonitrile | |
| Yellow | |
| C7H4FN | |
| 100 g | |
| p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene | |
| C1=CC(=CC=C1C#N)F | |
| 121.114 | |
| 121.11 | |
| Lumps |
| 1194-02-1 | |
| 188°C | |
| MFCD00001812 | |
| 1325 | |
| AEKVBBNGWBBYLL-UHFFFAOYSA-N | |
| 4-fluorobenzonitrile | |
| 14517 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 1194-02-1 | |
| 121.114 | |
| AEKVBBNGWBBYLL-UHFFFAOYSA-N | |
| 14517 | |
| C1=CC(=CC=C1C#N)F |
| C7H4FN | |
| MFCD00001812 | |
| p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene | |
| 4-fluorobenzonitrile |
Safety and Handling
RTECSNumber : DI4368500
TSCA : Yes