missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzonitrile 99.0+%, TCI America™

Catalog No. p-7129551
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F022125G 25 g
F0221100G 100 g
F0221500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F022125G Supplier TCI America Supplier No. F022125G
Only null left

Chemical Identifiers

CAS 1194-02-1
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.114
MDL Number MFCD00001812
InChI Key AEKVBBNGWBBYLL-UHFFFAOYSA-N
Synonym p-fluorobenzonitrile, benzonitrile, 4-fluoro, 4-cyanofluorobenzene, p-cyanofluorobenzene, benzonitrile, p-fluoro, 4-fluoro-benzonitrile, 4-fluorobenzenecarbonitrile, 4-fluorobenzonitril, pfbn, 1-cyano-4-fluorobenzene
PubChem CID 14517
IUPAC Name 4-fluorobenzonitrile
SMILES C1=CC(=CC=C1C#N)F

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 g
UN Number 1325
Formula Weight 121.11
Percent Purity ≥99.0% (GC)
Physical Form Lumps
Chemical Name or Material 4-Fluorobenzonitrile
RTECSNumber DI4368500
TSCA Yes