missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonamide 98.0+%, TCI America™

Catalog No. F09535G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09535G 5 g
F095325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09535G Supplier TCI America Supplier No. F09535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 402-46-0
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.177
MDL Number MFCD00025384
InChI Key LFLSATHZMYYIAQ-UHFFFAOYSA-N
Synonym p-fluorobenzenesulfonamide, 4-fluorobenzenesulphonamide, 4-fluorobenzene-1-sulfonamide, p-fluorobenzenesulphonamide, 4-flourobenzenesulfonamide, 4-fluoro-benzenesulfonamide, benzenesulfonamide, p-fluoro, benzenesulfonamide, 4-fluoro, pubchem2167, acmc-1acsj
PubChem CID 120231
IUPAC Name 4-fluorobenzenesulfonamide
SMILES C1=CC(=CC=C1F)S(=O)(=O)N

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 175.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzenesulfonamide
RTECSNumber DB2630000
TSCA No