accessibility menu, dialog, popup

UserName

NH2SOOF

CAS RN 402-46-0

IUPAC Name: 4-fluorobenzene-1-sulfonamide
Synonyms: 4-fluorobenzenesulfonamide 4-flourobenzenesulfonamide
Molecular Weight (g/mol): 175.18
Molecular Formula: C6H6FNO2S
InChi Key: LFLSATHZMYYIAQ-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)