missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonamide, 98+%

Catalog No. p-7037185
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55834MD 250 mg
AAH5583403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55834MD Supplier Thermo Scientific Chemicals Supplier No. H55834MD

CAS: 402-46-0 | C6H6FNO2S | 175.177 g/mol

4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzenesulfonamide, is used to produce bis-(4-fluoro-benzenesulfonyl)-amine with 4-fluoro-benzenesulfonyl chloride at temperature of 55 - 60. This reaction will need reagent OH-.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 402-46-0
Molecular Formula C6H6FNO2S
Molecular Weight (g/mol) 175.177
MDL Number MFCD00025384
InChI Key LFLSATHZMYYIAQ-UHFFFAOYSA-N
Synonym p-fluorobenzenesulfonamide, 4-fluorobenzenesulphonamide, 4-fluorobenzene-1-sulfonamide, p-fluorobenzenesulphonamide, 4-flourobenzenesulfonamide, 4-fluoro-benzenesulfonamide, benzenesulfonamide, p-fluoro, benzenesulfonamide, 4-fluoro, pubchem2167, acmc-1acsj
PubChem CID 120231
IUPAC Name 4-fluorobenzenesulfonamide
SMILES C1=CC(=CC=C1F)S(=O)(=O)N

Specifications

Melting Point 124°C to 127°C
Quantity 250 mg
UN Number UN2811
Solubility Information It is soluble in water.
Formula Weight 175.18
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzenesulfonamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-946-2
RTECSNumber DB2630000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only