missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzamide 98.0+%, TCI America™
Supplier: TCI America F027925G
Specifications
| 4-Fluorobenzamide | |
| 155°C | |
| C7H6FNO | |
| 25 g | |
| VNDHYTGVCGVETQ-UHFFFAOYSA-N | |
| 4-fluorobenzamide | |
| 71572 | |
| ≥98.0% (N) |
| 824-75-9 | |
| White-Yellow | |
| MFCD00007992 | |
| p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407 | |
| C1=CC(=CC=C1C(=O)N)F | |
| 139.129 | |
| 139.13 | |
| Crystalline Powder |
Chemical Identifiers
| 824-75-9 | |
| 139.129 | |
| VNDHYTGVCGVETQ-UHFFFAOYSA-N | |
| 71572 | |
| C1=CC(=CC=C1C(=O)N)F |
| C7H6FNO | |
| MFCD00007992 | |
| p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407 | |
| 4-fluorobenzamide |
Safety and Handling
RTECSNumber : CV4956666
TSCA : No