missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzamide 98.0+%, TCI America™

Catalog No. p-7129586
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
F02795G 5 g
F027925G 25 g
2 options

Catalog No. F02795G

Supplier: TCI America F02795G

Only null left

Chemical Identifiers

CAS 824-75-9
Molecular Formula C7H6FNO
Molecular Weight (g/mol) 139.129
MDL Number MFCD00007992
InChI Key VNDHYTGVCGVETQ-UHFFFAOYSA-N
Synonym p-fluorobenzamide, benzamide, 4-fluoro, benzamide, p-fluoro, p-fluorobenzoic acid amide, 4-fluoro-benzamide, unii-yb79f3xb1g, yb79f3xb1g, 4fluorobenzamide, 4-fluorobenzoamide, pubchem3407
PubChem CID 71572
IUPAC Name 4-fluorobenzamide
SMILES C1=CC(=CC=C1C(=O)N)F

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 139.13
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorobenzamide
RTECSNumber CV4956666
TSCA No