missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America F0191250ML
Specifications
| 4-Fluorobenzaldehyde | |
| Yellow | |
| C7H5FO | |
| 250 mL | |
| p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde | |
| C1=CC(=CC=C1C=O)F | |
| 124.114 | |
| 124.11 | |
| Liquid |
| 459-57-4 | |
| 181°C | |
| MFCD00003378 | |
| 1989 | |
| UOQXIWFBQSVDPP-UHFFFAOYSA-N | |
| 4-fluorobenzaldehyde | |
| 68023 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 459-57-4 | |
| 124.114 | |
| UOQXIWFBQSVDPP-UHFFFAOYSA-N | |
| 68023 | |
| C1=CC(=CC=C1C=O)F |
| C7H5FO | |
| MFCD00003378 | |
| p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde | |
| 4-fluorobenzaldehyde |
Safety and Handling
EINECSNumber : (3)-4458
TSCA : Yes