missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7129529
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F019125ML 25 mL
F0191250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F019125ML Supplier TCI America Supplier No. F019125ML
Only null left

Chemical Identifiers

CAS 459-57-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003378
InChI Key UOQXIWFBQSVDPP-UHFFFAOYSA-N
Synonym p-fluorobenzaldehyde, benzaldehyde, 4-fluoro, 4-fluoro-benzaldehyde, para-fluorobenzaldehyde, p-fluorbenzaldehyde, fluorobenzaldehyde 4-, benzaldehyde, p-fluoro, pfad, 4-fluoro benzaldehyde, para fluoro benzaldehyde
PubChem CID 68023
IUPAC Name 4-fluorobenzaldehyde
SMILES C1=CC(=CC=C1C=O)F

Specifications

Color Yellow
Boiling Point 181°C
Quantity 25 mL
UN Number 1989
Formula Weight 124.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluorobenzaldehyde
EINECSNumber (3)-4458
TSCA Yes