missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-o-phenylenediamine, 97%

CAS: 367-31-7 | C6H7FN2 | 126.134 g/mol

Supplier:  Thermo Scientific Chemicals A1397722

 View more versions of this product

Catalog No. AAA1397722


Only null left
Explore available promotions

Description

Description

4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

4-Fluoro-o-phenylenediamine
91°C to 95°C
MFCD00042228
UN2811
Light sensitive
Slightly soluble in water.
C1=CC(=C(C=C1F)N)N
126.134
126.13
367-31-7
C6H7FN2
100 g
2081075
1,2-diamino-4-fluorobenzene, 4-fluoro-1,2-phenylenediamine, 3,4-diaminofluorobenzene, 4-fluoro-o-phenylenediamine, 2-amino-4-fluoroaniline, 4-fluoro-1,2-diaminobenzene, 3,4-diamino-1-fluorobenzene, 5-fluoro-1,2-diaminobenzene, 4-fluoro-1,2-benzenediamine, 1,2-benzenediamine, 4-fluoro
KWEWNOOZQVJONF-UHFFFAOYSA-N
4-fluorobenzene-1,2-diamine
164584
97%

Chemical Identifiers

367-31-7
126.134
KWEWNOOZQVJONF-UHFFFAOYSA-N
164584
C1=CC(=C(C=C1F)N)N
C6H7FN2
MFCD00042228
1,2-diamino-4-fluorobenzene, 4-fluoro-1,2-phenylenediamine, 3,4-diaminofluorobenzene, 4-fluoro-o-phenylenediamine, 2-amino-4-fluoroaniline, 4-fluoro-1,2-diaminobenzene, 3,4-diamino-1-fluorobenzene, 5-fluoro-1,2-diaminobenzene, 4-fluoro-1,2-benzenediamine, 1,2-benzenediamine, 4-fluoro
4-fluorobenzene-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

H302+H332-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 206-691-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only