accessibility menu, dialog, popup

UserName

NH2FH2N

CAS RN 367-31-7

IUPAC Name: 4-fluorobenzene-1,2-diamine
Synonyms: 4-fluorobenzene-1,2-diamine
Molecular Weight (g/mol): 126.13
Molecular Formula: C6H7FN2
InChi Key: KWEWNOOZQVJONF-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1N
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)