Learn More
4-Fluoro-o-phenylenediamine, 97%
CAS: 367-31-7 | C6H7FN2 | 126.134 g/mol
Supplier: Thermo Scientific Chemicals A1397714
Description
4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
| 4-Fluoro-o-phenylenediamine | |
| 91°C to 95°C | |
| MFCD00042228 | |
| UN2811 | |
| Light sensitive | |
| Slightly soluble in water. | |
| C1=CC(=C(C=C1F)N)N | |
| 126.134 | |
| 126.13 |
| 367-31-7 | |
| C6H7FN2 | |
| 25 g | |
| 2081075 | |
| 1,2-diamino-4-fluorobenzene, 4-fluoro-1,2-phenylenediamine, 3,4-diaminofluorobenzene, 4-fluoro-o-phenylenediamine, 2-amino-4-fluoroaniline, 4-fluoro-1,2-diaminobenzene, 3,4-diamino-1-fluorobenzene, 5-fluoro-1,2-diaminobenzene, 4-fluoro-1,2-benzenediamine, 1,2-benzenediamine, 4-fluoro | |
| KWEWNOOZQVJONF-UHFFFAOYSA-N | |
| 4-fluorobenzene-1,2-diamine | |
| 164584 | |
| 97% |
Chemical Identifiers
| 367-31-7 | |
| 126.134 | |
| KWEWNOOZQVJONF-UHFFFAOYSA-N | |
| 164584 | |
| C1=CC(=C(C=C1F)N)N |
| C6H7FN2 | |
| MFCD00042228 | |
| 1,2-diamino-4-fluorobenzene, 4-fluoro-1,2-phenylenediamine, 3,4-diaminofluorobenzene, 4-fluoro-o-phenylenediamine, 2-amino-4-fluoroaniline, 4-fluoro-1,2-diaminobenzene, 3,4-diamino-1-fluorobenzene, 5-fluoro-1,2-diaminobenzene, 4-fluoro-1,2-benzenediamine, 1,2-benzenediamine, 4-fluoro | |
| 4-fluorobenzene-1,2-diamine |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
H302+H332-H311-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 206-691-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only