missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-methylbenzylamine 97.0+%, TCI America™

Catalog No. F08061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08061G 1 g
F08065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08061G Supplier TCI America Supplier No. F08061G

Chemical Identifiers

CAS 405-66-3
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03839848
InChI Key SZJIQLSCDIEJFC-UHFFFAOYSA-N
Synonym n-methyl-4-fluorobenzylamine, 4-fluoro-n-methylbenzylamine, 4-fluoro-benzyl-methyl-amine, 1-4-fluorophenyl-n-methylmethanamine, n-4-fluorobenzyl-n-methylamine, 4-fluoro-n-methyl benzylamine, 4-fluorophenyl methyl methyl amine, benzenemethanamine, 4-fluoro-n-methyl, 1-4-fluorophenyl-n-methyl-methanamine, acmc-1aek3
PubChem CID 736860
IUPAC Name 1-(4-fluorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 75°C
Quantity 1 g
UN Number 2735
Formula Weight 139.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-N-methylbenzylamine