accessibility menu, dialog, popup

UserName

CAS RN 405-66-3

IUPAC Name: [(4-fluorophenyl)methyl](methyl)azanium
Synonyms: [(4-fluorophenyl)methyl](methyl)azanium
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H11FN
InChi Key: SZJIQLSCDIEJFC-UHFFFAOYSA-O
SMILES: C[NH2+]CC1=CC=C(F)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)