missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-m-anisaldehyde 96.0+%, TCI America™

Catalog No. F04011G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04011G 1 g
F04015G 5 g
F040125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F04011G Supplier TCI America Supplier No. F04011G
Only null left

Chemical Identifiers

CAS 128495-46-5
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00143320
InChI Key NALVGTOMKSKFFV-UHFFFAOYSA-N
Synonym 3-methoxy-4-fluorobenzaldehyde, 4-fluoro-m-anisaldehyde, 4-fluoro-3-methoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-methoxy, pubchem1449, acmc-209bf6, ksc490m2d, 3-methoxy-4-fluoro benzaldehyde, 3-methoxy-4-fluoro-benzaldehyde, nalvgtomkskffv-uhfffaoysa
PubChem CID 2737358
IUPAC Name 4-fluoro-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1F

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-m-anisaldehyde