accessibility menu, dialog, popup

UserName

H3COOF

CAS RN 128495-46-5

IUPAC Name: 4-fluoro-3-methoxybenzaldehyde
Synonyms: 4-fluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: NALVGTOMKSKFFV-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC=C1F
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)