missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-alpha-methylbenzyl Alcohol 97.0+%, TCI America™
$205.05 - $809.00
Chemical Identifiers
| CAS | 403-41-8 |
|---|---|
| Molecular Formula | C8H9FO |
| Molecular Weight (g/mol) | 140.16 |
| MDL Number | MFCD00004515,MFCD03092997,MFCD03092998 |
| InChI Key | PSDSORRYQPTKSV-UHFFFAOYNA-N |
| Synonym | 1-4-fluorophenyl ethanol, 4-fluoro-alpha-methylbenzyl alcohol, 1-4-fluorophenyl ethan-1-ol, 1-p-fluorophenyl ethanol, p-fluorophenylmethylcarbinol, 4-fluorophenylmethylcarbinol, 1-4-fluorophenyl-1-ethanol, alpha-methyl-4-fluorobenzyl alcohol, 1-4-fluorophenyl-ethanol, 4-fluorophenyl methyl carbinol |
| PubChem CID | 73946 |
| IUPAC Name | 1-(4-fluorophenyl)ethan-1-ol |
| SMILES | CC(O)C1=CC=C(F)C=C1 |
Chemical Identifiers
| 403-41-8 | |
| 140.16 | |
| PSDSORRYQPTKSV-UHFFFAOYNA-N | |
| 73946 | |
| CC(O)C1=CC=C(F)C=C1 |
| C8H9FO | |
| MFCD00004515,MFCD03092997,MFCD03092998 | |
| 1-4-fluorophenyl ethanol, 4-fluoro-alpha-methylbenzyl alcohol, 1-4-fluorophenyl ethan-1-ol, 1-p-fluorophenyl ethanol, p-fluorophenylmethylcarbinol, 4-fluorophenylmethylcarbinol, 1-4-fluorophenyl-1-ethanol, alpha-methyl-4-fluorobenzyl alcohol, 1-4-fluorophenyl-ethanol, 4-fluorophenyl methyl carbinol | |
| 1-(4-fluorophenyl)ethan-1-ol |
Specifications
| 403-41-8 | |
| 123°C | |
| MFCD00004515,MFCD03092997,MFCD03092998 | |
| 1-4-fluorophenyl ethanol, 4-fluoro-alpha-methylbenzyl alcohol, 1-4-fluorophenyl ethan-1-ol, 1-p-fluorophenyl ethanol, p-fluorophenylmethylcarbinol, 4-fluorophenylmethylcarbinol, 1-4-fluorophenyl-1-ethanol, alpha-methyl-4-fluorobenzyl alcohol, 1-4-fluorophenyl-ethanol, 4-fluorophenyl methyl carbinol | |
| CC(O)C1=CC=C(F)C=C1 | |
| 140.16 | |
| 140.16 | |
| Liquid |
| Colorless | |
| C8H9FO | |
| 5 g | |
| PSDSORRYQPTKSV-UHFFFAOYNA-N | |
| 1-(4-fluorophenyl)ethan-1-ol | |
| 73946 | |
| ≥97.0% (GC) | |
| 4-Fluoro-alpha-methylbenzyl Alcohol |
Safety and Handling
TSCA : No