missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)phenol 98.0+%, TCI America™

Catalog No. F07545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07545G 5 g
F075425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07545G Supplier TCI America Supplier No. F07545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61721-07-1
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.102
MDL Number MFCD00043875
InChI Key DHPCRFYUUWAGAH-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl phenol, 2-fluoro-5-hydroxybenzotrifluoride, 3-trifluoromethyl-4-fluorophenol, phenol, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha-4-tetrafluoro-m-cresol, fhbtf, pubchem4147, acmc-209mvr, ksc495a2b
PubChem CID 601854
IUPAC Name 4-fluoro-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1O)C(F)(F)F)F

Specifications

Boiling Point 86°C
Quantity 5 g
UN Number 3265
Formula Weight 180.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)phenol
EINECSNumber (3)-4379
TSCA No