CAS RN 61721-07-1
IUPAC Name:
4-fluoro-3-(trifluoromethyl)phenol
Synonyms:
4-fluoro-3-(trifluoromethyl)phenol
Molecular Weight (g/mol):
180.1
Molecular Formula:
C7H4F4O
InChi Key:
DHPCRFYUUWAGAH-UHFFFAOYSA-N
SMILES:
OC1=CC=C(F)C(=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-(trifluoromethyl)phenol 98.0+%, TCI America™
CAS: 61721-07-1 Molecular Formula: C7H4F4O Molecular Weight (g/mol): 180.102 MDL Number: MFCD00043875 InChI Key: DHPCRFYUUWAGAH-UHFFFAOYSA-N Synonym: 4-fluoro-3-trifluoromethyl phenol,2-fluoro-5-hydroxybenzotrifluoride,3-trifluoromethyl-4-fluorophenol,phenol, 4-fluoro-3-trifluoromethyl,alpha,alpha,alpha-4-tetrafluoro-m-cresol,fhbtf,pubchem4147,acmc-209mvr,ksc495a2b PubChem CID: 601854 IUPAC Name: 4-fluoro-3-(trifluoromethyl)phenol SMILES: C1=CC(=C(C=C1O)C(F)(F)F)F