missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. F07605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07605G 5 g
F07601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07605G Supplier TCI America Supplier No. F07605G

Chemical Identifiers

CAS 67515-55-3
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00061294
InChI Key WZBPZYCJUADXRS-UHFFFAOYSA-M
Synonym 4-fluoro-3-trifluoromethyl benzoic acid, 4-fluoro-3-trifluomethyl benzoic acid, 4-fluoro-3-trifluoromethyl benzoicacid, alpha,alpha,alpha,4-tetrafluoro-m-toluic acid, benzoic acid, 4-fluoro-3-trifluoromethyl, pubchem4972, acmc-1bafh, ksc495k5d, rarechem al bo 0636
PubChem CID 522268
IUPAC Name 4-fluoro-3-(trifluoromethyl)benzoate
SMILES [O-]C(=O)C1=CC=C(F)C(=C1)C(F)(F)F

Specifications

Melting Point 115°C
Color White
Quantity 5 g
Formula Weight 208.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzoic Acid