CAS RN 67515-55-3
IUPAC Name:
4-fluoro-3-(trifluoromethyl)benzoate
Synonyms:
4-fluoro-3-(trifluoromethyl)benzoate
Molecular Weight (g/mol):
207.1
Molecular Formula:
C8H3F4O2
InChi Key:
WZBPZYCJUADXRS-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(F)C(=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
CAS: 67515-55-3 Molecular Formula: C8H3F4O2 Molecular Weight (g/mol): 207.10 MDL Number: MFCD00061294 InChI Key: WZBPZYCJUADXRS-UHFFFAOYSA-M Synonym: 4-fluoro-3-trifluoromethyl benzoic acid,4-fluoro-3-trifluomethyl benzoic acid,4-fluoro-3-trifluoromethyl benzoicacid,alpha,alpha,alpha,4-tetrafluoro-m-toluic acid,benzoic acid, 4-fluoro-3-trifluoromethyl,pubchem4972,acmc-1bafh,ksc495k5d,rarechem al bo 0636 PubChem CID: 522268 IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzoate SMILES: [O-]C(=O)C1=CC=C(F)C(=C1)C(F)(F)F