missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-phenoxybenzaldehyde 97.0+%, TCI America™

Catalog No. F05645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05645G 5 g
F056425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05645G Supplier TCI America Supplier No. F05645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68359-57-9
Molecular Formula C13H9FO2
Molecular Weight (g/mol) 216.21
MDL Number MFCD01318148
InChI Key JDICMOLUAHZVDS-UHFFFAOYSA-N
Synonym 4-fluoro-3-phenoxy-benzaldehyde, benzaldehyde, 4-fluoro-3-phenoxy, m-phenoxy-p-fluorobenzaldehyde, 2-fluoro-5-formyldiphenyl ether, 4-fluoro-3-phenoxy benzaldehyde, pubchem14130, 3-phenoxy-4fluorobenzaldehyde, ksc497q9b, 3-phenoxy-4-fluorobenzaldehyde, 3-phenoxy-4-fluoro-benzaldehyde
PubChem CID 110068
IUPAC Name 4-fluoro-3-phenoxybenzaldehyde
SMILES FC1=CC=C(C=O)C=C1OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 216.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-phenoxybenzaldehyde